Learn More
Agilent BioTek Wire Bottle Holder
Wire Bottle Holder for use with the 405 TS, 405 LS, ELx405 Select Deep Well, EL406 and 406 FX.
Brand:  Agilent BioTek 4032066
Additional Details : Weight : 1.36078kg
Product Code. 17814667
  • £56.65 / Each
Estimated Shipment: 07-06-2024
to see stock.
Description
Description
  • Holds two 4 L dispense or waste bottles
Specifications
Specifications
Bottle Stand
405 LS Microplate Washer, 405 TS Microplate Washer, ELx 405 Select Deep Well Microplate Washer, EL406 Microplate Washer, 406 FX Microplate Washer