missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI2 Mouse anti-Human, Mouse, Clone: 2A2, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | WIPI2 |
|---|---|
| Clone | 2A2 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, ELISA |
| Applications | Flow Cytometry, ELISA |
Description
WIPI2 Monoclonal specifically detects WIPI2 in Human, Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
| WIPI2 | |
| 0.2 mg/mL | |
| Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2 | |
| Recombinant fragment (around aa 418-436) of human WIPI2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| 2A2 | |
| Flow Cytometry 1 to 2 μg/million cells, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 26100 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title