missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93646-0.02ml
This item is not returnable.
View return policy
Description
WIPI2 Polyclonal antibody specifically detects WIPI2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| WIPI2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 187-436 of human WIPI2 (NP_057087.2). LAALAFDASGTKLATASEKGTVIRVFSIPEGQKLFEFRRGVKRCVSICSLAFSMDGMFLSASSNTETVHIFKLETVKEKPPEEPTTWTGYFGKVLMASTSYLPSQVTEMFNQGRAFATVRLPFCGHKNICSLATIQKIPRLLVGAADGYLYMYNLDPQEGGECALMKQHRLDGSLETTNEILDSASHDCPLVTQTYGAAAGKGTYVPSSPTRLAYTDDLGAVGGACLEDEASALRLDEDSEHPPMILRTD | |
| 0.02 mL | |
| Autophagy, Cell Biology, Endocrinology | |
| 26100 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction