missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI1 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82048AF488
This item is not returnable.
View return policy
Description
WIPI1 Polyclonal antibody specifically detects WIPI1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| WIPI1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 488 | |
| Atg18, Atg18 protein homolog, ATG18A, FLJ10055, WD repeat domain phosphoinositide-interacting protein 1, WD repeat domain, phosphoinositide interacting 1, WD40 repeat protein interacting with phosphoinositides of 49 kDa, WD40 repeat protein Interacting with phosphoInositides of 49kDa, WIPI 49 kDa, WIPI-1, WIPI-1 alpha, WIPI49ATG18 | |
| WIPI1 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human WIPI1. The immunogen is located within amino acids 350 - 400 of WIPI1. Amino Acid Squence: LGSGTTEENKENDLRPS | |
| 0.1 mL | |
| Autophagy, Cancer, Cell Biology, Endocrinology, Signal Transduction | |
| 55062 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction