missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WIPF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551995
This item is not returnable.
View return policy
Description
Immunogen sequence: PPPPVSRNGS TSRALPATPQ LPSRSGVDSP RSGPRPPLPP DRPSAGAPPP PPPSTSIRNG FQDSPCEDEW ESRFYFHPIS DLPPPEPYVQ TTKSYPSKLA RNESRSGSNR RERGAP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
This gene encodes a protein that plays an important role in the organization of the actin cytoskeleton. The encoded protein binds to a region of Wiskott-Aldrich syndrome protein that is frequently mutated in Wiskott-Aldrich syndrome, an X-linked recessive disorder. Impairment of the interaction between these two proteins may contribute to the disease. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
| WIPF1 | |
| Polyclonal | |
| Unconjugated | |
| WIPF1 | |
| AI115543; D2Ertd120e; MGC111041; Protein PRPL-2; PRPL-2; testicular tissue protein Li 226; WAS/WASL interacting protein family member 1; WAS/WASL interacting protein family, member 1; WAS/WASL-interacting protein family member 1; WAS2; WASP interacting protein; WASP-interacting protein; WASPIP; Wip; Wipf1; Wiskott-Aldrich syndrome protein interacting protein; wiskott-Aldrich syndrome protein-interacting protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7456 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43516 | |
| WIPF1 | |
| Recombinant protein corresponding to Human WIPF1. Recombinant protein control fragment (Product #RP-102120). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction