missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WIPF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570410
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.
This gene encodes a protein that plays an important role in the organization of the actin cytoskeleton. The encoded protein binds to a region of Wiskott-Aldrich syndrome protein that is frequently mutated in Wiskott-Aldrich syndrome, an X-linked recessive disorder. Impairment of the interaction between these two proteins may contribute to the disease. Two transcript variants encoding the same protein have been identified for this gene.
Specifications
| WIPF1 | |
| Polyclonal | |
| Unconjugated | |
| WIPF1 | |
| AI115543; D2Ertd120e; MGC111041; Protein PRPL-2; PRPL-2; testicular tissue protein Li 226; WAS/WASL interacting protein family member 1; WAS/WASL interacting protein family, member 1; WAS/WASL-interacting protein family member 1; WAS2; WASP interacting protein; WASP-interacting protein; WASPIP; Wip; Wipf1; Wiskott-Aldrich syndrome protein interacting protein; wiskott-Aldrich syndrome protein-interacting protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 117538 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6IN36 | |
| WIPF1 | |
| synthetic peptide directed towards the N-terminal region of rat WIPF1. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction