missing translation for 'onlineSavingsMsg'
Learn More

WIPF1 Polyclonal Antibody

Product Code. 15963775
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15963775 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15963775 Supplier Invitrogen Supplier No. PA577268

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

WIPF1 Polyclonal Antibody

WIPF1 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen WIPF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene WIPF1
Gene Accession No. O43516, Q6IN36, Q8K1I7
Gene Alias AI115543; D2Ertd120e; MGC111041; Protein PRPL-2; PRPL-2; testicular tissue protein Li 226; WAS/WASL interacting protein family member 1; WAS/WASL interacting protein family, member 1; WAS/WASL-interacting protein family member 1; WAS2; WASP interacting protein; WASP-interacting protein; WASPIP; Wip; Wipf1; Wiskott-Aldrich syndrome protein interacting protein; wiskott-Aldrich syndrome protein-interacting protein
Gene Symbols WIPF1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 431-480 of Human WIPF1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117538, 215280, 7456
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.