missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Winter Pro Gloves
Description
- Resistant to cold temperatures and chemicals.
- Watertight.
- Suitable for use in wet/oily environments.
- PVC glove on 100% seamless, woven, knitted cotton for comfort and flexibility.
- Patterned finish.
- Designed with a cuff for forearm protection.
Specifications
Specifications
| Certifications/Compliance | EC Category PPE 3, EC Certificate Number 0075/797/162/03/13/0204, EC Attestation Number 0075/797/162/03/13/0204, CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 374/2-2003 Chemical hazards, EN 511-2003 Cold hazards |
| Classification | Dexterity: 5 |
| Coating Material | Cotton, PVC |
| Color | Blue |
| Cut Resistance Rating | 2 |
| Material | PVC |
| Product Type | Gloves |
| Size | 8 |
| Thickness (Metric) | 1.4 mm |
| For Use With (Application) | Building, Construction, Chemical, Fishing, Mining, Quarrying, Petrochemical and Water Treatment industries. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?