missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Premiere Ecoforce™ Window and Glass Cleaner RTU
Click to view available options
Volume (Metric):
750 mL
Description
- Fast and effective
- Leaves no smears, just clean and shiny surfaces that sparkle
- Non-abrasive making it ideal for mirrors, chrome and glass
- Ready to use trigger spray bottle
Specifications
Specifications
| Product Type | Window and Glass Cleaner |
| Concentration | RTU |
| For Use With (Application) | Non abrasive making it ideal for mirrors,chrome and glass |
| Quantity | 6/Cs. |
| Volume (Metric) | 750 mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction