missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Thin Wall Quartz EPR Sample Tube

Manufactured from naturally occurring quartz with the highest purity to minimize the background signal. Bel-Art SP Scienceware™ Thin Wall Quartz EPR Sample Tube may contain Fe2O3 which is paramagnetic (1200 ppm) and has certain impact on shimming quality depending on the magnetic field strength.

Brand:  Bel-Art™ 66000-0015

Code : 03

Additional Details : Weight : 0.27900kg

 View more versions of this product

Product Code. 15378578

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Tight ID and OD tolerances ensure 100% compatibility to spectrometers
  • Individually wrapped to prevent contamination
  • Recommended for experiments with small organic molecule (Molecular Weight ∼500) at ambient temperature only
Specifications

Specifications

Sample Tube
250 mm
4 mm
710-SQ-250M
Quartz
5 mm
0.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers