Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Spinner Bearing
Product Code. 15368608 Shop All SP Scienceware Products
Click to view available options
Model:
SB-5-7
SB-8-7
Diameter (Metric) Outer:
For tubes 5mm O.D.
For tubes 8mm O.D.
Unit Size:
Each
Product Code. 15368608
Brand: Bel-Art™ 660000042
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ensure shimming quality of your glass tubes. Bel-Art SP Scienceware™ Wilmad™ Spinner Bearing will determine which of your NMR tubes are warped and bent.
  • Useful in labs that wash, dry and reuse precision NMR tubes
  • Warped tubes will bind in the spinner bearing while good tubes spin freely
  • Keep a tube tester beside every NMR spectrometer for easy access
Specifications
For Use With (Equipment) Quartz, Supracil, or Precision tubes
Diameter (Metric) Outer For tubes 5mm O.D.
Model SB-5-7