Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Pressure Vacuum Valve NMR Tube
Opens by turning counter-clockwise. Bel-Art SP Scienceware™ Wilmad™ Pressure Vacuum Valve NMR Tube is the most reliable NMR tube on the market for medium range pressure and ultra-vacuum experiments.
Brand: Bel-Art™ 66304-0571
Code : 03
Additional Details : Weight : 0.04310kg
Alert:
Due to the nature of glass, extreme caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.
Description
- Connects to a 1/8" (3.175mm) metal vacuum line using Swagelok™ fittings
- Pressure vacuum valve is made of PTFE, all other parts are Pyrex™ or equivalent
Specifications
51/25 µm | |
1/8in. (3.175mm) stainless steel or brass vacuum line | |
5 mm | |
0.38 mm |
PTFE, Pyrex™ | |
178 mm | |
7 in. | |
507-PV-7 |