missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ Open Cap for 4 to 5 mm Tubes
Description
- Comes complete with open top screw cap
- Inert to most organic solvents and compounds but poor to strongly corrosive materials
- Remains reliable after multiple punctures
Specifications
Specifications
| Product Type | Open Cap Replacement |
| Description | Model: TR-OC-01 |
| Color | Black |
| Quantity | 12 pieces |
| For Use With (Equipment) | 4 and 5 mm Screw Cap NMR Tubes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?