missing translation for 'onlineSavingsMsg'
Learn More

Wilmad™ SP Scienceware™ 12mm O.D. Thin Walled Precision NMR Tube

To maximize SNR, Precision NMR Tubes have minimal paramagnetic impurities that would impact shimming. Tight ID and OD tolerance as small as 0.0065mm accommodates Wilmad inserts

Brand:  Wilmad™ 66060-1285

Code : 03

Additional Details : Weight : 0.04310kg

 View more versions of this product

Product Code. 15368748

  • £105.49 / Pack of 5


Call for latest availability
+44 (0)1509 555 500
Add to basket

Alert:

Caps are not recommended for use with samples dissolved in chloroform-d or acetone-d6.


Specifications

Specifications

203.2mm
11.050±0.013mm
For experiments requiring critical shimming quality (high/ultrahigh field, non-spinning, multi-dimensional, multi-nuclei, DNP experiments and studies involving biological samples)
514A-1PP-8-5
12.065±0.008mm
60MHz
0.51mm
Videos