missing translation for 'onlineSavingsMsg'
Learn More

Wilforlide A, MedChemExpress

Product Code. 16481418
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481418 5 mg 5mg
16438998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481418 Supplier Medchem Express Supplier No. HYN0476

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Wilforlide A is a bioactive triterpene isolated from Tripterygium wilfordii Hook f. Wilforlide A has anti-inflammatory and immune suppressive effects.

Wilforlide A is a bioactive triterpene isolated from Tripterygium wilfordii Hook f. Wilforlide A has anti-inflammatory and immune suppressive effects.

Specifications

Chemical Name or Material Wilforlide A
CAS 84104-71-2
Color White
Purity Grade Notes Research
Molecular Formula C30H46O3
Quantity 5 mg
Synonym Regelide Abruslactone A
Solubility Information DMSO : 1 mg/mL (2.20 mM; Need ultrasonic)
SMILES C[C@@]1(C2=O)C[C@]([H])(O2)[C@]3(C)CC[C@@]4(C)[C@]5(C)CC[C@@]6([H])C(C)(C)[C@@H](O)CC[C@]6(C)[C@@]5([H])CC=C4[C@]3([H])C1
Molecular Weight (g/mol) 454.68
Formula Weight 454.68
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.