missing translation for 'onlineSavingsMsg'
Learn More

Wilforine, MedChemExpress

Product Code. 16429178
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429178 10 mg 10mg
16421588 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16429178 Supplier Medchem Express Supplier No. HYN0899
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Wilforine is a sesquiterpene pyridine alkaloid; important bioactive compound in T.

Wilforine is a sesquiterpene pyridine alkaloid

Specifications

Chemical Name or Material Wilforine
CAS 11088-09-8
Color White
Purity Grade Notes Research
Molecular Formula C43H49NO18
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (57.61 mM; Need ultrasonic)
SMILES CC(OCC(C1OC(C)=O)(C(C(OC(C2=CC=CC=C2)=O)C3OC(C4C)=O)OC(C)=O)C(OC(C)(COC(C5=CC=CN=C5CC4)=O)C6C1OC(C)=O)(C3(O)C)C6OC(C)=O)=O
Molecular Weight (g/mol) 867.85
Formula Weight 867.85
Percent Purity 98.03%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.