missing translation for 'onlineSavingsMsg'
Learn More

WIKI4, MedChemExpress

Product Code. 16453227
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453227 50 mg 50mg
16447208 10 mg 10mg
16439737 5 mg 5mg
16413556 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16453227 Supplier Medchem Express Supplier No. HY16910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WIKI4 is a potent tankyrase inhibitor with an IC50 of 26 nM for TNKS2. WIKI4 potently inhibits Wnt/β-catenin signaling and that its half-maximal response dose is 75 nM. WIKI4 mediates its effects on Wnt/β-catenin signaling by inhibiting the enzymatic activity of TNKS2. WIKI4 is cytotoxic to SCLC cells with an IC50 value of 0.02 μM.

WIKI4 is a potent tankyrase inhibitor with an IC50 of 26 nM for TNKS2. WIKI4 potently inhibits Wnt/β-catenin signaling and that its half-maximal response dose is 75 nM. WIKI4 mediates its effects on Wnt/β-catenin signaling by inhibiting the enzymatic activity of TNKS2. WIKI4 is cytotoxic to SCLC cells with an IC50 value of 0.02 μM.

Specifications

Chemical Name or Material WIKI4
CAS 838818-26-1
Color Tan
Purity Grade Notes Research
Molecular Formula C29H23N5O3S
Quantity 50 mg
Solubility Information DMSO : 6.8 mg/mL (13.04 mM; Need ultrasonic and warming)
SMILES O=C1N(CCCSC2=NN=C(C3=CC=NC=C3)N2C4=CC=C(OC)C=C4)C(C5=CC=CC6=CC=CC1=C56)=O
Molecular Weight (g/mol) 521.59
Formula Weight 521.59
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.