missing translation for 'onlineSavingsMsg'
Learn More

WIKI4, MedChemExpress

Product Code. 16413556
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413556

Brand: Medchem Express HY16910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WIKI4 is a potent tankyrase inhibitor with an IC50 of 26 nM for TNKS2. WIKI4 potently inhibits Wnt/β-catenin signaling and that its half-maximal response dose is 75 nM. WIKI4 mediates its effects on Wnt/β-catenin signaling by inhibiting the enzymatic activity of TNKS2. WIKI4 is cytotoxic to SCLC cells with an IC50 value of 0.02 μM.

WIKI4 is a potent tankyrase inhibitor with an IC50 of 26 nM for TNKS2. WIKI4 potently inhibits Wnt/β-catenin signaling and that its half-maximal response dose is 75 nM. WIKI4 mediates its effects on Wnt/β-catenin signaling by inhibiting the enzymatic activity of TNKS2. WIKI4 is cytotoxic to SCLC cells with an IC50 value of 0.02 μM.

Specifications

Chemical Name or Material WIKI4
CAS 838818-26-1
Color Tan
Purity Grade Notes Research
Molecular Formula C29H23N5O3S
Quantity 25 mg
Solubility Information DMSO : 6.8 mg/mL (13.04 mM; Need ultrasonic and warming)
SMILES O=C1N(CCCSC2=NN=C(C3=CC=NC=C3)N2C4=CC=C(OC)C=C4)C(C5=CC=CC6=CC=CC1=C56)=O
Molecular Weight (g/mol) 521.59
Formula Weight 521.59
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.