missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF-1 Rabbit anti-Human, Mouse, Rat, Clone: 1X6C9, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £395.00
Specifications
| Antigen | WIF-1 |
|---|---|
| Clone | 1X6C9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18309465
|
Novus Biologicals
NBP3-16580-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18329364
|
Novus Biologicals
NBP3-16580-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| WIF-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction, Wnt Signaling Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 11197 | |
| IgG | |
| Affinity purified |
| 1X6C9 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| WIF-1, WNT inhibitory factor 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 280-379 of human WIF-1 (Q9Y5W5). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title