missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF-1 Antibody (RM0144-3M51), PE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22520PE
This item is not returnable.
View return policy
Description
WIF-1 Monoclonal antibody specifically detects WIF-1 in Mouse samples. It is validated for Western Blot
Specifications
| WIF-1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| RM0144-3M51 | |
| Western Blot | |
| WIF-1, WNT inhibitory factor 1 | |
| Mouse recombinant protein of WIF-1 | |
| 0.1 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 11197 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction