missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIF-1 Antibody (RM0144-3M51), Alexa Fluor™ 750, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22520AF750
This item is not returnable.
View return policy
Description
WIF-1 Monoclonal specifically detects WIF-1 in Mouse samples. It is validated for Western Blot.
Specifications
| WIF-1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| WIF1 | |
| Mouse recombinant protein of WIF-1 | |
| 0.1 mL | |
| 11197 | |
| Mouse | |
| IgG2 |
| Western Blot | |
| RM0144-3M51 | |
| Western Blot | |
| WIF-1, WNT inhibitory factor 1 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| This was produced from a hybridoma (mouse myeloma) fused with spleen cells from a rat immunized with mouse recombinant protein of WIF-1. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction