Learn More
Motic™ Widefield Eyepiece WF20X/10mm
Click to view available options
Magnification Power:
20X
Unit Size:
Each
Description
- Widefield style provides improved field of view and greater eye relief.
- Magnification: 20X.
- Field of view: 10mm.
Specifications
Specifications
| Type | Widefield |
| Description | FOV: 10mm |
| For Use With (Equipment) | ST-39, SFC-11, SMZ-140 Microscopes |
| Magnification Power | 20X |