Learn More
Motic™ Widefield Eyepiece N-WF15X/F.N.16 with Diopter Adjustment (ESD)
Product Code. 15455522
Click to view available options
Magnification Power:
15X
Unit Size:
Each
Product Code. 15455522
Brand: Motic™ 1101001402731
Only null left
Add to basket
Add to basket
Widen your field of view for more complete specimen viewing. Motic™ Widefield Eyepiece N-WF15X/16mm with Diopter Adjustment (ESD) provides an improved field of view and greater eye relief.
  • Widefield style and diopter adjustment provides improved field of view and greater eye relief.
  • Magnification: 10X.
  • Field number: 16.
Specifications
Type Widefield
Description F.N.: 16
For Use With (Equipment) SM-171, GM Series Microscopes
Includes Diopter adjustment, ESD electrostatic shock protection
Magnification Power 15X