missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ WIC1
GREENER_CHOICE

Product Code. 18737016
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18737016

Brand: Tocris Bioscience™ 7580/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of Wnt/β-catenin signaling

WIC1 is an inhibitor of the canonical Wnt/β-catenin signaling pathway. In vitro, it reduces ABSC (Airway Basal Stem Cell) proliferation and induces ciliated epithelial cell differentiation, in a model of squamous lung cancer. WIC1 also represses the phosphorylation of β-cateninY489 and Tp63 expression.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[4-(4-Ethyl-1-piperazinyl)phenyl]-2-oxo-2 H-1-benzopyran-3-carboxamide
CAS 943083-58-7
Quantity 10 mg
Target beta-Catenin
Molecular Formula C22H23N3O3
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.