missing translation for 'onlineSavingsMsg'
Learn More

WIBG Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Product Code. 18358556
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18358556 100 μg 100µL
18356612 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18358556 Supplier Novus Biologicals Supplier No. NBP315586100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WIBG Polyclonal antibody specifically detects WIBG in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen WIBG
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Formulation PBS, 50% glycerol, pH7.3
Gene Alias MGC13064, partner of Y14 and mago, PYMProtein wibg homolog, within bgcn homolog (Drosophila)
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-204 of human WIBG (NP_115721.1). MEAAGSPAATETGKYIASTQRPDGTWRKQRRVKEGYVPQEEVPVYENKYVKFFKSKPELPPGLSPEATAPVTPSRPEGGEPGLSKTAKRNLKRKEKRRQQQEKGEAEALSRTLDKVSLEETAQLPSAPQGSRAAPTAASDQPDSAATTEKAKKIKNLKKKLRQVEELQQRIQAGEVSQPSKEQLEKLARRRALEEELEDLELGL
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 84305
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.