missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIBG Antibody (OTI5E2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74868CL1
This item is not returnable.
View return policy
Description
WIBG Monoclonal antibody specifically detects WIBG in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| WIBG | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC13064, partner of Y14 and mago, PYMProtein wibg homolog, within bgcn homolog (Drosophila) | |
| Full length human recombinant protein of human WIBG (NP_115721) produced in HEK293T cell. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 84305 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI5E2 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction