missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WHSC2 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-05975G
This item is not returnable.
View return policy
Description
WHSC2 Polyclonal antibody specifically detects WHSC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| WHSC2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FLJ10442, negative elongation factor A, NELFA, NELF-AFLJ25112, Wolf-Hirschhorn syndrome candidate 2, Wolf-Hirschhorn syndrome candidate 2 protein | |
| The immunogen recognized by this antibody maps to a region between residue 499 and 549 of human negative elongation factor A using the numbering given in entry BAB18651.1 (GeneID 7469). | |
| 0.1 mL | |
| Cancer | |
| 7469 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction