missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Whole Cell Stains
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 × 96 Tests
Unit Size:
Each
Beskrivning
Provide excellent staining for high-content screening assays and other fluorescent cellular imaging methods involving fluorescence microscopy using Thermo Scientific™ Whole Cell Stains.
These stains are intense, highly photostable and match the output wavelengths of common fluorescence instrumentation. Effective image analysis in cell-based assays and fluorescence microscopy of the localization of specific targets requires fluorescent labeling of the entire cells, nuclei or major cellular components. Whole cell stains identify cell numbers, cell borders and cell morphologies as useful primary objects for assessment of specific protein targets and their redistribution in response to treatment conditions.
- Provide excellent staining for HCA and fluorescence microscopy
- Intense, highly photostable and match the output wavelengths of common fluorescence instrumentation
- When the whole cell is the primary object used to identify and count individual cells, effectively distinguish intact cells from bordering cells (with analysis by appropriate software)
Includes:
Fluorescent stains to detect whole cells (blue, green, orange or red)
Requires:
Fluorescence microscope, Thermo Scientific™ Cellomics™ ArrayScan™ VTI or other HCS reader
Specifikationer
Specifikationer
| Description | Cellomics Whole Cell Stain Green |
| Quantity | 5 × 96 Tests |
| Target | Whole cell (green); Ex/Em = 493/518nm |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering