missing translation for 'onlineSavingsMsg'
Learn More

White Spirit v.p. For Laboratory Use, Chem Lab

Product Code. 12155660
Change view
Click to view available options
Quantity:
5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12155660 5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 12155660 Supplier Chem-Lab Supplier No. CL00.2302.5000
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Specifications

CAS 8052-41-3
Boiling Range 150 to 196 °C
Color Colorless
Density 0.78 g/mL
Flash Point 36 °C
Packaging Polyethylene
Molecular Formula C8H15BrO2
Quantity 5 L
InChI Key IMSPZLCTPSXORJ-UHFFFAOYSA-N
SMILES CC(=O)OCCCCCCBr
IUPAC Name 6-bromohexyl acetate
Molecular Weight (g/mol) 223.11
Grade Very Pure
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • White Spirit v.p.
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: white spirit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.