Learn More
Honeywell™ White Range Super Mont Blanc S2 Shoes
Product Code. 12621323
Click to view available options
Size:
38
39
40
42
44
47
Unit Size:
Each
Product Code. 12621323
Brand: Honeywell™ 624074847/7
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ White Range Super Mont Blanc S2 Shoes are a slip-on ankle boot with a 200J toe cap.
  • Offers optimal protection
  • White treated leather upper with limited stitching
  • Resistant to liquid projections and easy to clean
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance European Directive: 89/686/EEC - EC PPE Category II - Standard: EN 345-1 - Marking : S2 Quality Assurance: ISO 9001:2008
Color White
Material Leather
Size 47
Sole Features White, double density, injected, perforated
Sole Material Polyurethane
Toe Style Metallic 200J Toe Cap
Product Type Safety Footwear