Learn More
SHOWA™ White General Purpose Gloves, Model B0610
Product Code. p-7229370
Click to view available options
Size:
6
7
8
9
Unit Size:
10 pairs
Product Code. 17897381
Brand: SHOWA™ BO61006S
Only null left
Add to basket
Add to basket
This light, elastic low-lint general-purpose glove is resistant to deformation so it keeps its shape for longer.
  • Designed for optimum freedom of movement, the B0610 has a short wrist cuff.
  • The uncoated, seamless nylon liner reduces irritation and allergy risks while maintaining dexterity and tactility.
Specifications
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Nylon
Product Type Glove
Size 6
Sterility Non-sterile
Style Seamless Knit
FDA 510 (k) No
For Use With (Application) Mechanical and engineering, Automotive repairs and maintenance, Integrated circuits, Intricate parts handling, Microelectronic, Semiconductors, Cleanrooms, Quality control
Glove Cuff Style Elasticated
Series General Purpose