missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coverguard™ White Coverall 5M20
Description
- CE PPE Cat. III
- Limited protection against chemical liquid splash
- Protects against chemical solid particles in the air
- Anti-static protection
- White
- 55 g/m2 SPP + microporous film fabric
- Central zip with adhesive flap
- 3 pans elasticated hood
- Elasticated: waist, wrist with elastic thumbloop, ankle tightening, slingbar
- Self-adhesive chin-strap
Specifications
Specifications
| Size | Medium |
| Gender | Unisex |
| Material | PPE |
| Color | White |
| Ankle Style | Elastic |
| Cuff Style | Thumb Loop |
| Hood Type | Elastic with Self-adhesive Chin Strap |
| Closure Type | Central Zip with Adhesive Flap |
| Sterility | Non-sterile |
| Coating | Microporous Film |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?