missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coverguard™ White Coverall 5M20
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Pakningsstørrelse:
Pack of 25
Beskrivelse
- CE PPE Cat. III
- Limited protection against chemical liquid splash
- Protects against chemical solid particles in the air
- Anti-static protection
- White
- 55 g/m2 SPP + microporous film fabric
- Central zip with adhesive flap
- 3 pans elasticated hood
- Elasticated: waist, wrist with elastic thumbloop, ankle tightening, slingbar
- Self-adhesive chin-strap
Tekniske data
Tekniske data
| Size | Small |
| Gender | Unisex |
| Material | PPE |
| Color | White |
| Ankle Style | Elastic |
| Cuff Style | Thumb Loop |
| Hood Type | Elastic with Self-adhesive Chin Strap |
| Closure Type | Central Zip with Adhesive Flap |
| Sterility | Non-sterile |
| Coating | Microporous Film |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion