missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Socorex™ White Capillary Tubes
Click to view available options
Volume (Metric):
1 to 5 μL
5 to 25 μL
Unit Size:
Pack of 200
Description
- White
- Replacement glass capillaries
- For Acura™ 846 positive displacement pipettes
- Color coded to match corresponding pipette
Specifications
Specifications
| Volume (Metric) | 1 to 5 μL |
| Sterility | Sterile |
| Color | White |
| Quantity | 200/Pk. |
| Product Type | Capillary Tube |
| Disposable | Single-use |
| For Use With (Equipment) | Acura 846 Positive Displacement Pipette |
| Material | Glass |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction