missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ WHIP Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB110-61626PE
This item is not returnable.
View return policy
Description
WHIP Polyclonal antibody specifically detects WHIP in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| WHIP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 56897 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| PE | |
| ATPase WRNIP1, bA420G6.2, EC 3.6.1, FLJ22526, putative helicase RUVBL, Werner helicase interacting protein 1, Werner helicase-interacting protein 1, WHIPRP11-420G6.2 | |
| The immunogen recognized by this antibody maps to a region between residue 400 and 450 of human Werner helicase interacting protein 1 using the numbering given in entry NP_064520.2 (GeneID 56897). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction