missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WHIP Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-61626UV
This item is not returnable.
View return policy
Description
WHIP Polyclonal specifically detects WHIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| WHIP | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| ATPase WRNIP1, bA420G6.2, EC 3.6.1, FLJ22526, putative helicase RUVBL, Werner helicase interacting protein 1, Werner helicase-interacting protein 1, WHIPRP11-420G6.2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56897 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| WRNIP1 | |
| The immunogen recognized by this antibody maps to a region between residue 400 and 450 of human Werner helicase interacting protein 1 using the numbering given in entry NP_064520.2 (GeneID 56897). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction