missing translation for 'onlineSavingsMsg'
Learn More

WHIP Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18297075 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18297075 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18297075 Supplier Novus Biologicals Supplier No. NB11061626AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WHIP Polyclonal specifically detects WHIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WHIP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ATPase WRNIP1, bA420G6.2, EC 3.6.1, FLJ22526, putative helicase RUVBL, Werner helicase interacting protein 1, Werner helicase-interacting protein 1, WHIPRP11-420G6.2
Gene Symbols WRNIP1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 400 and 450 of human Werner helicase interacting protein 1 using the numbering given in entry NP_064520.2 (GeneID 56897).
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 56897
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.