missing translation for 'onlineSavingsMsg'
Learn More

WHI-P180, MedChemExpress

Product Code. 16406818
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16406818

Brand: Medchem Express HY15769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WHI-P180 (Janex 3) is a multi-kinase inhibitor; inhibits RET, KDR and EGFR with IC50s of 5 nM, 66 nM and 4 μM, respectively.

WHI-P180 (Janex 3) is a multi-kinase inhibitor

Specifications

Chemical Name or Material WHI-P180
CAS 211555-08-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H15N3O3
Quantity 10 mg
Synonym Janex 3
Solubility Information DMSO : 25 mg/mL (84.09 mM; Need ultrasonic)
SMILES COC1=CC2=NC=NC(NC3=CC=CC(O)=C3)=C2C=C1OC
Molecular Weight (g/mol) 297.31
Formula Weight 297.31
Percent Purity 99.62%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.