missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies WHI-P131
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- WHI-P131 is an inhibitor of Janus kinase 3 (JAK3) with IC50 values of 9 and 78 μM against human and mouse proteins, respectively (Sudbeck et al.) It has also been reported to show significant inhibition of other kinases, including epidermal growth factor receptor (EGFR) in the nanomolar range (Changelian et al.; Uckun et al.) No significant inhibition of JAK1 or JAK2 has been observed (Sudbeck et al.)
- Induces apoptosis and cell death in human glioblastoma cell lines U373 and U87 (Narla et al.)
- Inhibits lipopolysaccharide (LPS)-induced nitric oxide synthase expression and nitric oxide production in macrophages (Sareila et al.)
- Inhibits proliferation of neural stem cell-containing neurospheres at lower concentrations than the concentration required to inhibit proliferation of astrocyte cultures (Diamandis et al.)
Specifications
Specifications
| CAS | 202475-60-3 |
| Synonym | Janex-1 |
| Quantity | 1 mg |
| Molecular Formula | C16H15N3O3 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction