missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WHI-P 154
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
WHI-P 154 is a JAK3 inhibitor (IC50 = 1.8 μM) that displays no activity at JAK1 or JAK2. Inhibits STAT1 activation, iNOS expression and NO production in macrophages in vitro. Also inhibits other common kinases including EGFR (IC50 = 4 nM), Src, Abl, VEGFR, MAPK and PI 3-K and induces apoptosis in human glioblastoma cell lines (IC50 = 158 μM). Induces differentiation of neural progenitor cells.
Specifications
Specifications
| Chemical Name or Material | 2-Bromo-4-[(6,7-dimethoxy-4-quinazolinyl)amino]phenol |
| CAS | 211555-04-3 |
| Assay Percent Range | >99% |
| Quantity | 5 mg |
| Molecular Formula | C16H14BrN3O3 |
| Formula Weight | Observed MW: 376.2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction