missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Screw Caps for Glass Bottles and Vials

Product Code. 11718899 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
15-425
18-400
24-400
Liner Material:
PTFE/Polyethylene
PTFE/Silicone
Polyethylene Foam
Polyvinyl
No. per Case:
144/Cs.
200/Cs.
Unit Size:
Pack of 144
Pack of 200
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Liner Material No. per Case unitSize
11718899 15-425 Polyethylene Foam 144/Cs. Pack of 144
11794199 24-400 PTFE/Silicone 200/Cs. Pack of 200
11728899 18-400 Polyethylene Foam 144/Cs. Pack of 144
11788819 18-400 Polyvinyl 144/Cs. Pack of 144
11758809 18-400 PTFE/Polyethylene 144/Cs. Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11718899 Supplier DWK Life Sciences Supplier No. 239274

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

With clear surface for writing

  • Available with different liners (PTFE/silicone, foamed polyethylene, poly-vinyl, PTFE faced foamed polyethylene)
  • With choice of open or closed top

Caps with PTFE/silicone liner:

  • Provides heat resistance
  • Glass filled
  • Suited for EPA samples
Caps with foamed polyethylene liner:
  • Chemical resistant
  • Tight seal
Caps with poly-vinyl liner:
  • With moisture barrier
  • Suited for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Caps with PTFE faced foamed polyethylene liner:
  • Chemical resistant
  • Provides compressibility and sealing properties
  • Suited for high purity chemicals, strong acids and solvents

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 144/Cs.
Liner Material Polyethylene Foam
Neck Finish 15-425
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.