missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Screw Caps with bonded PTFE Faced Silicone Liner for E-Z Ex-Traction™ Vials
Shop All Wheaton Science Products ProductsDescription
- Autoclavable
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 100 Cs. |
| No. per Pack | 100/Cs. |
| Neck Finish | 24-400 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | 20mL Vial |
| Closure Liner | PTFE/Silicone |
| Autoclavable | Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?