missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials

Product Code. 10725373 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
13-425
15-425
22-400
24-400
No. per Case:
100 Cs.
1000 Cs.
250 Cs.
500 Cs.
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Pack of 500
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish No. per Case unitSize
10725373 13-425 250 Cs. Pack of 250
10526863 13-425 1000 Cs. Pack of 1000
10048272 15-425 250 Cs. Pack of 250
10139822 15-425 1000 Cs. Pack of 1000
10556863 22-400 100 Cs. Pack of 100
10119922 22-400 500 Cs. Pack of 500
10785553 24-400 100 Cs. Pack of 100
10557833 24-400 500 Cs. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10725373 Supplier DWK Life Sciences Supplier No. W240840

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Vea la política de devoluciones

Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

  • Autoclavable

TRUSTED_SUSTAINABILITY

Especificaciones

Color White
Material Polypropylene
No. per Case 250 Cs.
No. per Pack 250/Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Closure Liner PTFE/Silicone
Autoclavable Autoclavable
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.