missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners

Product Code. 11724229 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Liner Material:
PTFE/Polyethylene
Polyethylene Foam
Material:
Polypropylene
Polypropylene, PTFE
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Unit Size:
Pack of 144
Pack of 48
Pack of 72
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Liner Material Material No. per Case unitSize
11724229 33-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11784219 20-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11714229 28-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11708819 63-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
11798809 53-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11704229 24-400 PTFE/Polyethylene Polypropylene, PTFE 144 Cs. Pack of 144
11778809 43-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11788809 45-400 Polyethylene Foam Polypropylene 72 Cs. Pack of 72
11794219 22-400 Polyethylene Foam Polypropylene 144 Cs. Pack of 144
11734229 48-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11744229 58-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11768809 38-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
10113052 89-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
11754229 70-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 11724229 Supplier DWK Life Sciences Supplier No. 239236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Excellent for environmental samples, pharmaceuticals and diagnostic reagents

  • PTFE faced foamed polyethylene liner offers chemical resistance
  • Polyethylene foam provides compressibility and sealing properties
  • Good for high purity chemicals, strong acids, and solvents
  • For usewith glass bottles or vials
  • Polypropylene
  • Screw cap
  • White

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 144 Cs.
Liner Material PTFE/Polyethylene
Neck Finish 33-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.