missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners

Product Code. 11704229 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Liner Material:
PTFE/Polyethylene
Polyethylene Foam
Material:
Polypropylene
Polypropylene, PTFE
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Unit Size:
Pack of 144
Pack of 48
Pack of 72
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Liner Material Material No. per Case unitSize
11704229 24-400 PTFE/Polyethylene Polypropylene, PTFE 144 Cs. Pack of 144
11784219 20-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11714229 28-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11708819 63-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
11798809 53-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11778809 43-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11788809 45-400 Polyethylene Foam Polypropylene 72 Cs. Pack of 72
11724229 33-400 PTFE/Polyethylene Polypropylene 144 Cs. Pack of 144
11794219 22-400 Polyethylene Foam Polypropylene 144 Cs. Pack of 144
11734229 48-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11744229 58-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
11768809 38-400 PTFE/Polyethylene Polypropylene 72 Cs. Pack of 72
10113052 89-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
11754229 70-400 PTFE/Polyethylene Polypropylene 48 Cs. Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11704229 Fornitore DWK Life Sciences N. del fornitore 239233
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Excellent for environmental samples, pharmaceuticals and diagnostic reagents

  • PTFE faced foamed polyethylene liner offers chemical resistance
  • Polyethylene foam provides compressibility and sealing properties
  • Good for high purity chemicals, strong acids, and solvents
  • For usewith glass bottles or vials
  • Polypropylene
  • Screw cap
  • White

TRUSTED_SUSTAINABILITY

Specifica

Color White
Material Polypropylene, PTFE
No. per Case 144 Cs.
Liner Material PTFE/Polyethylene
Neck Finish 24-400
Certifications/Compliance ISO 9001
Product Type Screw Cap
For Use With (Equipment) Glass bottle or vial
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.