missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners
Shop All Wheaton Science Products ProductsDescription
- White polypropylene screw cap provides clear surface for writing on cap
- Poly-vinyl liner provides an excellent moisture barrier
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 144 Cs. |
| Neck Finish | 24-400 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Polypropylene Cap |
| For Use With (Equipment) | Round bottle |
| Closure Liner | Polyvinyl |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?