missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners

Product Code. 11708829 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
22-400
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
89-400
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Unit Size:
Pack of 144
Pack of 48
Pack of 72
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish No. per Case unitSize
11708829 22-400 144 Cs. Pack of 144
10296624 70-400 48 Cs. Pack of 48
11778829 48-400 72 Cs. Pack of 72
11718829 24-400 144 Cs. Pack of 144
11748829 38-400 72 Cs. Pack of 72
11798829 58-400 72 Cs. Pack of 72
10546653 89-400 48 Cs. Pack of 48
11758829 43-400 72 Cs. Pack of 72
11728829 28-400 144 Cs. Pack of 144
11788829 53-400 72 Cs. Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 11708829 Supplier DWK Life Sciences Supplier No. 239207
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products

  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 144 Cs.
Neck Finish 22-400
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Round bottle
Closure Liner Polyvinyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.