missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners

Product Code. 11798899 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
13-425
24-400
28-400
38-400
43-400
48-400
53-400
58-400
70-400
Material:
Glass
Polypropylene
No. per Case:
144 Cs.
48 Cs.
72 Cs.
Unit Size:
Pack of 144
Pack of 48
Pack of 72
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish Material No. per Case unitSize
11798899 43-400 Polypropylene 72 Cs. Pack of 72
11738909 58-400 Polypropylene 72 Cs. Pack of 72
11718909 48-400 Polypropylene 72 Cs. Pack of 72
11708899 13-425 Polypropylene 144 Cs. Pack of 144
11728909 53-400 Glass 72 Cs. Pack of 72
11758899 24-400 Polypropylene 144 Cs. Pack of 144
11758909 70-400 Polypropylene 48 Cs. Pack of 48
11768899 28-400 Polypropylene 144 Cs. Pack of 144
11788899 38-400 Polypropylene 72 Cs. Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11798899 Supplier DWK Life Sciences Supplier No. 239286

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

  • Liner provides tight seal
  • Liner Material: Foam, Polyethylene
  • Polypropylene
  • Screw Cap
  • White

Use these caps with glass bottles or vials

Order Info

Each pack contains 72

TRUSTED_SUSTAINABILITY

Specifications

Color White
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 43-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Glass bottle or vial
Closure Liner Polyethylene Foam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.