Learn More
DWK Life Sciences Wheaton™ Tamper-Evident Caps
Product Code. 11704319 Shop All Wheaton Science Products Products
Change view
Click to view available options
No. per Case:
2500 Case
5000 Case
Unit Size:
100g
Pack of 5000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. per Case unitSize
11704319 2500 Case 100g
11714319 5000 Case Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11704319 Supplier DWK Life Sciences Supplier No. W224198
Contents cannot be accessed without removing cap or leaving evidence of puncture or insert
  • Use with Wheaton Uni-Dose Bottles and Vials
  • Three-piece Pulp / PVDC / PE lined aluminum caps have unique metal inserts
  • A metal tab with a rolled safety edge “perks up” to provide visual proof of tight seal
  • Caps remove quickly and easily in one piece
  • Cat. No. 03-084-130 is stamped inside “For Oral Use”
  • Attach using Wheaton Crimper
TRUSTED_SUSTAINABILITY
Specifications
Includes Rolled safety edge
Material Aluminum
No. per Case 2500 Case
Liner Material Pulp/PVDC/Polyethylene
Certifications/Compliance ISO 9001
For Use With (Equipment) Uni dose bottle and vial