missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Ultra Pure Straight Plug Stoppers

Product Code. 11708959 Shop All Wheaton Science Products Products
Change view
Click to view available options
Diameter (Metric) Bottom:
7 mm
13 mm
Diameter (Metric) Top:
13 mm
20 mm
Unit Size:
Pack of 1000
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Bottom Diameter (Metric) Top unitSize
11708959 13 mm 20 mm Pack of 1000
11778949 7 mm 13 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11708959 Supplier DWK Life Sciences Supplier No. W224100405
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides for extremely high chemical purity and low gas permeability. Wheaton™ Ultra Pure Straight Plug Stoppers are manufactured from an ultra pure bromobutyl formulation.

  • Autoclavable
  • Low level of extractables
  • Complies with US, European and Japanese Pharmacopeias
  • Free of latex, nitrosamines and 2-MCBT (2-mercaptobenzothiazole)
  • Compatible with applications such as Water for Injection (WFI) where rubber can cause problems
TRUSTED_SUSTAINABILITY

Specifications

Hardness 47 Durometer
Autoclavable Autoclavable
Color Gray
Diameter (Metric) Bottom 13 mm
Diameter (Metric) Top 20 mm
Material Bromobutyl Rubber
Product Type Straight Plug Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.